Tweezers, Straight Fine Tip, Style 3C, Excelta Corp®
63042-128
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- Descripción:Tweezers, very fine point, 2 star quality
- Matriz:Stainless steel 316L, anti-magnetic
- Curvature:Straight
- Referencia:63042-128
- Longitud total:106.25 mm (4.25")
- Nº de proveedor:3C-SA-PI
Style 3C is ideal for precision work, including under magnification.
- Ideal for working under magnification
- Available in three different materials
- Available in three different qualtiy grades
- Ergonomic handles are standard option
- Teflon coated tip option for non-marring
The 'Style 3C' series straight very fine precision pointed tweezer is designed for work under magnification. The overall length is 4.25".The different options offered are ergonomic handles or teflon coated tips (to prevent scratching).depending on your application. We offer 3 different qualities. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic).