ContactUsLinkComponent
Tweezers, Curved Tapered Relieved Tip, Style 5A, Excelta Corp®
Tweezers, Curved Tapered Relieved Tip, Style 5A, Excelta Corp®
Catalog # 63042-678
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Tweezers, Curved Tapered Relieved Tip, Style 5A, Excelta Corp®
Catalog # 63042-678
Supplier Number:  5B-SA
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Descripción:
    Tweezers, ultra fine point, 3 star quality
  • Matriz:
    Stainless steel 316L, anti-magnetic
  • Curvature:
    45° bent
  • Referencia:
    63042-678
  • Longitud total:
    106.25 mm (4.25")
  • Nº de proveedor:
    5B-SA
  • Tip Width:
    0.004" (0.1 mm)
  • Tip Length:
    0.05 mm (0.002")
Specifications
About this item
These are ideal for microscopy, featuring offset tapered tip for excellent visibility.
  • Tapered offset tips for greater visibility/accessibility
  • Very fine points for precision
  • Offered in 3 different materials
  • Three quality levels
  • Available with ergonomic grips
These angled/bent tapered ultra-fine pointed tweezer ares primarily used for microscopy. The offset angle offers easy access and is designed for good visibility. We offer 3 different qualities. The higher the quality, the more precise the tips. They are offered in stainless steel (rust-resistant) or stainless anti-magnetic steel. The 'ET' version features our Ergo-Tweeze® soft cushioned grips (10¹⁰ ohms/sq surface resistivity), which also protect the user from heat.