Eisco Glass Test Tubes with Screw Cap
Catalog # 470323-180
Supplier: EISCO SCIENTIFIC LLC
Specifications
- Capacidad:15 ml
- Referencia:470323-180
- O.D.×L:16×125 mm
- Bottom Style:Round bottom
Specifications
About this item
High quality glass test tubes with bakelite screw cap and rubber liner for general laboratory use.
- Made from high quality Borosilicate 3.3 glass
- Autoclavable
- Superior chemical durability
- Low temperature gradient- can withstand direct flame, high temperatures and thermal shocks
Eisco Labs Borosilicate glass has the following heat tolerances: strain point –515 °C; annealing point 565 °C; softening point 820 °C. They manufacture their own products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Delivery: Bakelite screw cap with rubber liner included.