VWR® Maximum Protection Sterile Mask with Ear Loops
Catalog # 76420-852
Supplier: VWR International
Specifications
- Esterilidad:Sterile
- Color:White
- Descripción:Cleanroom face mask with ear loops
- Referencia:76420-852
Specifications
About this item
The new sterile ear loop mask expands upon the VWR® mask types in the popular cleanroom Maximum Protection line by providing a sterile option.
- 3-ply masks
- Sterile 10⁻⁶, gamma sterilization
- Ear loop configuration
- Natural, latex-free
- Low particle shedding
VWR® Maximum Protection sterile masks provide high bacterial and particle filtration efficiency, while remaining breathable and fluid-resistant. Malleable noseband ensures a secure seal.
Suitable for use in cleanroom environments, production, and research.
Packaging: Masks are packaged 10 per bag, 5 inner bags per outer bag, 500 masks per case.