HALO® RP-Amide, HPLC Columns, Advanced Materials Technology
Catalog # 76443-906
Supplier: Advanced Materials Technology
Specifications
- Formato de columna:Analytical
- Ø int.:2.10 mm
- Tamaño de partícula:2 µm
- Tamaño de poro:90 Å
- Longitud:100 mm
- Extremo tapado:Yes
- Código USP:L60
- Rango de pH:2 - 9
- Temperature limit:60 ⁰C (pH 2); 40 ⁰C (pH 9)
- Column Hardware:Acero inoxidable
- Referencia:76443-906
- Phase:RP- Amide
Specifications
About this item
HALO® RP-Amide columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high-resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å RP-Amide columns are optimized for high performance in HPLC, UHPLC and LCMS small molecule applications.
- Ideal for basic compounds (alcohols, acids, phenols, catechins)
- Alternate selectivity to alkyl phases
- 100% aqueous compatible
- Particle sizes: 2, 2.7 or 5 μm superficially porous particle silica ('solid-core', 'core-shell')
HALO® guard columns are available for this phase.