ContactUsLinkComponent
HotBlock® Watch Glasses and Reflux Caps, Environmental Express®
HotBlock® Watch Glasses and Reflux Caps, Environmental Express®
  76437-120
undefined
HotBlock® Watch Glasses and Reflux Caps, Environmental Express®
  76437-120
 :  SC506
  • Descripción:
    Disposable reflux caps, PP, for 50 ml cups
  • Referencia:
    76437-120
Choose from reflux caps or watch glasses compatible with digestion cups.
  • Optimize use of your HotBlock® digestion cup
These disposable 34 mm reflux caps fit onto our 50 ml digestion cups. This special cap has a 12.7 mm hole in its center to allow for the addition of reagents during digestion.
The watch glasses feature a ribbed design which allows partial reflux during digestion. Watch glass has a large center stem to aid in handling.