Edge® 48-706 Heavy-Duty Cut Resistant Industrial Gloves, Ansell
Catalog # 76424-318
Supplier: ANSELL HEALTHCARE PRODUCTS LLC MX
Specifications
- Color del guante:Blue,Green
- Tamaño del guante:9
- Latex/No contiene Latex:Latex
- Con polvo/sin polvo:Sin polvo
- Longitud:8.66 - 10.63" (220 - 270 mm)
- Tipo de puño:Puño elástico
- Calibre:13
- Acabado:Palm coated
- Referencia:76424-318
Specifications
About this item
Cost effective solution for medium weight applications requiring a high level of cut performance in dry or slightly oily environments.
- High durability Nitrile foam coating
- Excellent comfort and dexterity
- High level of cut performance EN ISO level C cut performance (ANSI Level 3)
- Excellent abrasion resistance
- Breathable liner provides a high level of comfort for end users
- Good level of grip in dry and slightly oily environments
Made of a seamless liner, palm dipped into foam nitrile coating. ANSI cut level A3 and EN ISO 13997 rated level C provide a good level of cut resistance. The nitrile coating delivers very good performance in terms of abrasion resistance and grip. The best alternative against heavy-duty leather and polyurethane palm dipped cut resistant knitted gloves. Use primarily for automotive, construction, glass manufacturing, machinery/equipment, and metal fabrication applications.
Certifications: EN 388:2016. EN 420:2003 + A1:2009. ANSI ABR 5. ANSI Cut A3. Category II. REACH Compliant.
Packaging: Gloves are packaged 12 pairs per bag, 12 bags per carton.