Orders LinkContactUsLinkComponent
FlaskScrubber® Glassware Washers, Labconco®
Freestanding FlaskScrubber glassware washer, 115 V, 50/60 Hz, 1 phase, 16 A, 104.3 kg (230 lbs.), 617×706×818 - 919 mm (24.3×27.8×32.2 - 36.2")
  76420-720
   Waste Reducing|Water Efficient
 :  412001000
FlaskScrubber® Glassware Washers, Labconco®
  76420-720
   Waste Reducing|Water Efficient
FlaskScrubber® Glassware Washers, Labconco®
FlaskScrubber® Glassware Washers, Labconco®
Freestanding FlaskScrubber glassware washer, 115 V, 50/60 Hz, 1 phase, 16 A, 104.3 kg (230 lbs.), 617×706×818 - 919 mm (24.3×27.8×32.2 - 36.2")
$327,410.00
Each (1UN)
Versatile and efficient glassware washers designed for efficient and consistent washing of narrow-neck glassware and general labware.
  • Touch screen display with intuitive CleanWorks OS software
  • Flexible design allows for use of spindle injection and standard racks
  • Maximum wash temperature of 93 °C removes tough contaminants
  • Advanced wash pump maximizes water circulation up to 112 gal/min (424 liters/min)
  • Standard forced air spindle drying with SpeedVent technology
  • Durable Type 304 stainless steel chamber and racks
These glassware washers are designed for efficient and consistent washing of narrow-neck glassware and general purpose labware. The CleanWorks OS software is accessible through a full-color touch screen interface and provides intuitive and clear monitoring of wash loads – ensuring repeatable, effective and traceable cleaning.
FlaskScrubber models reach a maximum of 93 °C wash temperature and quickly dry glassware with standard forced air spindle injection drying with SpeedVent heat-management technology. These models include a 36-place lower spindle rack, and may be outfitted with a wide variety inserts and additional racks to accommodate all types of labware.
 : ETL, cETL, and CE listed. Compliant with UL61010, CAN/CSA C22.2, CE.
 : Two year warranty on parts and labor. Optional accessories include various inserts for specific glassware types, detergents, spindle and standard racks, labware carts, and water purifiers.
 : Lower spindle rack included.
 : All single phase models require hard wiring to a 20 amp dedicated circuit. All three phase models require hard wiring to a 30 amp dedicated circuit.
  • Description:
    Freestanding FlaskScrubber glassware washer
  • W×D×H:
    617×706×818 - 919 mm (24.3×27.8×32.2 - 36.2")
  • Electrical:
    115 V, 50/60 Hz, 1 phase, 16 A
  • Shipping Weight:
    104.3 kg (230 lbs.)
  • Cat. No.:
    76420-720
  • Environmentally Preferable:
    Waste Reducing|Water Efficient