Orders LinkContactUsLinkComponent
VWR® Flat Caps for Screw Cap Microtubes
VWR® Flat Caps for Screw Cap Microtubes
Clear, Non sterile
Catalog # 76417-216
Supplier:   VWR International
VWR® Flat Caps for Screw Cap Microtubes
Catalog # 76417-216
Supplier:  VWR International
VWR® Flat Caps for Screw Cap Microtubes
VWR® Flat Caps for Screw Cap Microtubes
Clear, Non sterile
Price
$1,077.70
Log in Para disponibilidad
Paquete de 500 Case de 10 (500UN)
Product Details & Documents
Flat caps with O-ring for use with VWR® screw cap microtubes only (series 76417-xxx).
Feature ³/₄ turn to easily open and close tubes requiring less revolutions than competitor tubes.
Especificaciones
  • Sterility:
    Non sterile
  • Cap Color:
    Clear