Test Strips, Water Hardness, Dosatest, VWR Chemicals BDH®
Catálogo # BDH85442.601
Proveedor: VWR International
Número CAS:
Especificaciones
- Descripción:Hardness 5 - 25 °d
- Referencia:BDH85442.601
- Rango:< 3 · > 5 · > 10 · > 15 · > 20 · > 25
- No. de tiras:100
Especificaciones
Acerca de este artículo
A ready-to-use kit that fits into your lab coat pocket.
- Rapid and convenient
- Easy to handle
- Choice of ranges for required precision
Ideal to monitor water hardness, especially suitable for boiler testing, aquaria and laundries. Semi-quantitative results can be obtained between 10 and 120 seconds.
The color charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
1 mmol/litre =10 °f = 5.60 °d= 7,022 °e
1 mmol/litre= 100.09 mg/l CaCO₃
Embalaje: Container with 100 test strips 6 x 95 mm