Orders LinkContactUsLinkComponent
Objectives for REVOLVE Microscopes, Echo
Supplier:   DISCOVER ECHO INC.
Objectives for REVOLVE Microscopes, Echo
Supplier:  DISCOVER ECHO INC.
Certificates
Objectives for REVOLVE Microscopes, Echo
2 Options Available Below
Product Details & Documents
Product Family Options
Información del producto
  • Magnification
    Working Distance
    Aperture
    Availability
    Price
  • 75992-490
    Fluorite phase objective
    Fluorite phase objective
    17 mm
    0.13 NA
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN)
    Supplier Number: JMAG0043
  • 75990-662
    Fluorite ELWD phase objective
    Fluorite ELWD phase objective
    40×
    3.0 - 4.2 mm
    0.60 NA
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN)
    Supplier Number: JMAG0404
  • 76490-274
    Apochromat objective
    Apochromat objective
    40×
    0.18 mm
    0.95 NA
    Discontinued
    Supplier Number: JMAG9402