Horseshoe Magnet
7.5 cm
Catalog # 470012-762
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: SHM030
Jump to:
Product Details & Documents
Strong steel magnets, zinc-plated with North and South poles marked.
- Keeper included
Supplied with keeper, 3 inches long.
Specifications
- Description:7.5 cm