Ceramic Disk Magnet
Magnet disk, 5 mm, 25 mm
Catalog # 470003-343
Supplier: DOWLING MAGNETS,LLC
Número de proveedor #: MC06
Jump to:
Product Details & Documents
Ceramic disc magnet, 25×5 mm.
Especificaciones
- Description:Magnet disk
- Height:5 mm
- Weight:25 mm