Pony Beads
Pony beads, assorted colors, 1 lb package
Catalog # 470309-222
Supplier: THE BEADERY
Número de proveedor #: 750SV076
Jump to:
Product Details & Documents
Plastic pony beads are perfect for a huge variety of crafts and experiments.
- 9 mm Diameter
- Assorted Colors
- For Ages 6+
Colored beads are a great tool for teachers and parents. Students can learn to string, sort colors, weigh and count, all while having fun and being creative. Bring out the fun in the classroom, scouts, birthday parties and on a rainy day.
: WARNING: CHOKING HAZARD. Children under 8 years can choke or suffocate. Adult supervision is required.
Especificaciones
- Description:Pony beads, assorted colors, 1 lb package