Orders LinkContactUsLinkComponent
Eisco® Wax Lined Dissection Tray
Eisco® Wax Lined Dissection Tray
Aluminum dissecting tray with clear wax, 10.6×7×1.5" (27×18×4 cm)
Catalog # 470199-842
Supplier:   VWR International
Eisco® Wax Lined Dissection Tray
Catalog # 470199-842
Supplier:  VWR International
Eisco® Wax Lined Dissection Tray
Eisco® Wax Lined Dissection Tray
Aluminum dissecting tray with clear wax, 10.6×7×1.5" (27×18×4 cm)
Price
$450.39
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
High-quality stainless steel dissection tray with wax liner for pinning specimens.
  • Premium stainless steel
  • Wax liner for pinning specimens
Perfect for entomology and dissection labs. The wax liner allows pinning of specimens for greater agility and observation. Stainless steel is durable and easy to clean. The approximate 2" depth helps contain liquids.
Especificaciones
  • Description:
    Aluminum dissecting tray with clear wax
  • L×W×D:
    10.6×7×1.5" (27×18×4 cm)