VWR® Irradiated PE Cleanroom Tapes
Catalog # 76311-238
Supplier: VWR International
Specifications
- Anchura:1.9 cm (0.75" )
- Color:Transparent
- Descripción:Polyethylene tape, irradiated
- Longitud:33 m (36 yds.)
- Matriz:Polyethylene film, acrylic adhesive
- Referencia:76311-238
Specifications
About this item
An irradiated polyethylene tape with low particulate and bio-burden counts for use in critical and aseptic cleanroom applications. With low residual residue on plastics and other smooth surfaces in the cleanroom, this tape is ideal for plate sealing and surface marking.
- Irradiated, pressure-sensitive polyethylene tape
- Low particulate and bio-burden counts
- Low outgassing
- Ideal for sealing, color-coding, and marking in aseptic area
- Processed in a cleanroom
- Individually double bagged
- Available in a range of widths
- 3" plastic core
- Roll length: 33 m (36 yard)
Processed in a controlled environment, wound on a plastic core, and individually double bagged. Each roll is 36 yards and available in 0.75, 1, or 2" widths. White or transparent finishes are offered.
Delivery: Supplied with full certification of irradiation and lot traceability.