VWR® Wire Racks
Supplier: VWR International
Jump to:
Acerca de este artículo
These HDPE coated wire racks are durable for transport or sample handling.
- Chemically-resistant
- Racks can be used between temperatures of –40 °C and 121 °C
- Slotted bottoms
- Holds all brands and types of tubes/bottles for each size range
- Square wells and three deck levels help keep tubes secure
- Autoclavable
Suitable for conical, round or flat-bottom glass or plastic tubes and bottles.
9 Options Available Below
Product Details & Documents
These HDPE coated wire racks are durable for transport or sample handling.
- Chemically-resistant
- Racks can be used between temperatures of –40 °C and 121 °C
- Slotted bottoms
- Holds all brands and types of tubes/bottles for each size range
- Square wells and three deck levels help keep tubes secure
- Autoclavable
Suitable for conical, round or flat-bottom glass or plastic tubes and bottles.
Documents
Certificates
Buscar por número de loteProduct Family Options
Información del producto
- Hole ØColorNo. of HolesL×W×HAvailabilityPrice
- 10 - 13 mmBlue72 (6×12)204.6×105×65 mmEach (1UN)$334.94Case de 48 (1UN)$365.07
- 16 - 20 mmBlue40 (4×10)240.4×98.8×80 mmEach (1UN)$488.49Case de 48 (1UN)$532.45
- 16 - 20 mmBlue80 (8×10)240.4×193.2×80 mmEach (1UN)$946.72Case de 24 (1UN)$1,031.92
- 20 - 24 mmBlue40 (4×10)260.4×106.8×80 mmEach (1UN)$907.60Case de 24 (1UN)$989.29
- 22 - 25 mmBlue40 (4×10)290.4×118.8×80 mmEach (1UN)$946.72Case de 24 (1UN)$1,031.92
- 20 - 25 mmBlue80 (8×10)290.4×233.2×80 mmEach (1UN)$907.60Case de 12 (1UN)$989.29
- 25 - 30 mmBlue48 (4×12)276.8×209.6×98 mmEach (1UN)$1,037.32Case de 8 (1UN)$1,130.68
- 30 - 40 mmBlue24 (3×8)356.8×138.8×98 mmEach (1UN)$688.73Case de 12 (1UN)$750.72
- 30 - 40 mmBlue48 (4×12)356.8×269.6×98 mmEach (1UN)$1,292.81Case de 8 (1UN)$1,409.17