Wire Gauzes, Plain
Supplier: UNITED SCIENTIFIC SUPPLIES
Jump to:
About this item
Sturdy wire mesh for supporting glassware over a Bunsen burner.
Long lasting, heat- and tear-resistant iron
- Available in a variety of sizes
Sheet of cross-hatched wire used to support glassware during heating. Galvanized iron wire provides a long-life expectancy and is resistant to tearing over time. Edges are folded over for a clean perimeter surface.
8 Options Available Below
Product Details & Documents
Sturdy wire mesh for supporting glassware over a Bunsen burner.
Long lasting, heat- and tear-resistant iron
- Available in a variety of sizes
Sheet of cross-hatched wire used to support glassware during heating. Galvanized iron wire provides a long-life expectancy and is resistant to tearing over time. Edges are folded over for a clean perimeter surface.
Product Family Options
Product Information
- SizeAvailabilityPrice
- 470019-498Wire Gauze, Plain, 4" x 4"Supplier Number: WGPL44-PK/12
- 470144-820Wire Gauze, Plain, 4" x 4"Wire Gauze, Plain, 4" x 4"10×10 cm (4×4")Each (1UN)$20.06Case de 10 (1UN)$21.86Supplier Number: WGPL44
- 470176-062Wire Gauze, Plain, 4" x 4"Wire Gauze, Plain, 4" x 4"10×10 cm (4×4")Each (1UN)$20.06Paquete de 10 (1UN)$21.86Supplier Number: WGPL44
- 470019-500Wire Gauze, Plain, 5" x 5"Supplier Number: WGPL55-PK/12
- 470144-818Wire Gauze, Plain, 5" x 5"Wire Gauze, Plain, 5" x 5"13×13 cm (5×5")Each (1UN)$33.05Case de 10 (1UN)$36.02Supplier Number: WGPL55
- 470188-996Wire Gauze, Plain, 5" x 5"Wire Gauze, Plain, 5" x 5"13×13 cm (5×5")Each (1UN)$33.05Paquete de 10 (1UN)$36.02Supplier Number: WGPL55
- 470019-502Wire Gauze, Plain, 6" x 6"Supplier Number: WGPL66-PK/12
- 470188-998Wire Gauze, Plain, 6" x 6"Wire Gauze, Plain, 6" x 6"15×15 cm (6×6")Each (1UN)$41.20Paquete de 10 (1UN)$44.91Supplier Number: WGPL66