Avantor® ACE® Phenyl, HPLC columns, 10 µm
Catalog # 76389-090
Supplier: Avantor
Specifications
- Formato de columna:Analytical
- Ø int.:3.90 mm
- Tamaño de partícula:100 µm
- Tamaño de poro:100 Å
- Presión máx.:400 bar(5801.51 psi)
- Longitud:300 mm
- Extremo tapado:Yes
- Código USP:L11
- Rango de pH:2 - 8
- Surface area:300 m²/g
- Carga de carbono:9.5%
- Referencia:76389-090
- Phase:Phenyl
Specifications
About this item
These ultra inert Avantor® ACE® Phenyl columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Offers strong π-π interactions
- Ideal for analytes with π-bonding, electron withdrawing or delocalised groups
- Wide range of particle sizes (2, 3, 5, 10 µm) and complementary phases complementary phases
- Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.