Avantor® ACE® Generix® C8 & C8(2), Analytical HPLC Columns
Catalog # 76395-046
Supplier: Avantor
Specifications
- Formato de columna:Analytical
- Ø int.:4.60 mm
- Tamaño de partícula:5 µm
- Tamaño de poro:95 Å
- Longitud:150 mm
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:ISO 14001:2015 (530840 UM15)
- Extremo tapado:Yes
- Código USP:L7
- Rango de pH:2 - 8
- Surface area:350 m²/g
- Carga de carbono:11.9%
- Referencia:76395-046
- Phase:C8(2)
Specifications
About this item
These high performance HPLC columns contain high purity spherical silica.
- Ultra-narrow particle size distribution for excellent column efficiency
- Sharp, highly symmetrical peaks for acidic, basic and neutral analytes
- Excellent column to column and batch to batch reproducibility
- These columns are made of stainless steel
Each column is individually tested and supplied with a test chromatogram.
Two generations of Avantor® ACE® Generix® C8 exist: C8 and C8(2) they differ in pore size, carbon load and surface area.