Avantor® Partisil ODS3, HPLC Columns
Supplier: Avantor
Certificates
About this item
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
- Easy to use
- Irregular porous silica
Avantor® Partisil® ODS3 is an intermediate carbon load C18 phase used for pharmaceutical, natural products, food, biological and environmental pollutants applications.
Specifications
- Catalog No:
- 76402-130
- 76402-104
- 76401-876
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)