Avantor® Partisil PAC, HPLC Columns
Catalog # 76401-920
Supplier: Avantor
Specifications
- Ø int.:4.60 mm
- Tamaño de partícula:10 µm
- Tamaño de poro:85 Å
- Longitud:250 mm
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:ISO 14001:2015 (530840 UM15)
- Referencia:76401-920
- Phase:PAC
Specifications
About this item
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
- Easy to use
- Irregular porous silica
- Stainless steel column hardware
Avantor® Partisil® PAC is a polar amino cyano bonded phase with secondary amine groups for good thermal and chemical stability. Fast equilibration enables multiple separation mechanisms, including adsorption, reversed-phase and weak anion-exchange, to be used. The phase is particularly suitable for carbohydrate separations.
Caution: * Use with freestanding holder HI-161 and column coupler HI-081, please see 'HPLC Guard Column Holders and Accessories'.