Avantor® Partisil SAX, HPLC Columns
Catalog # 76401-958
Supplier: Avantor
Specifications
- Ø int.:4.60 mm
- Tamaño de partícula:10 µm
- Tamaño de poro:85 Å
- Longitud:250 mm
- Extremo tapado:No
- Rango de pH:1.5 - 7.5
- Carga de carbono:10%
- Referencia:76401-958
- Phase:SAX
Specifications
About this item
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
- Easy to use
- Irregular porous silica
- High efficiency
- Stainless steel column hardware
Avantor® Partisil® SAX is a strong anion exchange phase based on quaternary ammonium groups and is stable over the pH range 1.5 to 7.5. At intermediate pH, use of a Solvecon eluent conditioning column is recommended to further enhance column lifetime. Suitable for use with nucleic acids, organic acids and inorganic anions.