Avantor® Ultrasphere® Octyl (C8), HPLC Columns
Supplier: Avantor
Acerca de este artículo
Avantor® Ultrasphere® columns remain widely referenced within both industry and academia and are recognized to provide excellent chromatographic performance.
- 3 and 5 μm spherical porous silica
- High reproducibility, narrow particle size distribution
- Optimal surface coverage for long column lifetime
2 Options Available Below
Product Details & Documents
Avantor® Ultrasphere® columns remain widely referenced within both industry and academia and are recognized to provide excellent chromatographic performance.
- 3 and 5 μm spherical porous silica
- High reproducibility, narrow particle size distribution
- Optimal surface coverage for long column lifetime
Specifications for Product Family
Especificaciones
- Catalog No:
- 76398-090
- 76398-088
- Endcapped:
- Yes
- Yes
Especificaciones
Product Family Options
Información del producto
- Column FormatPhasePore sizeParticle SizeØ int.LengthPacking MaterialAvailabilityPrice
- Supplier Number: 235333Especificaciones:
More
Specifications
Cat. No.76398-090EndcappedYes - Supplier Number: 235332Especificaciones:
More
Specifications
Cat. No.76398-088EndcappedYes
Accessories
Información del producto
- Catalog #AvailabilityPrice
- Avantor® Ultrasphere® XL, column holder assembly, stainless steelAvantor® Ultrasphere® XL, column holder assembly, stainless steel76398-102Log in Para disponibilidadEach (1UN) - $4,445.62