Avantor® Partisil Silica, HPLC Columns
76402-044
: Avantor
- Formato de columna:Analytical
- Ø int.:4.60 mm
- Tamaño de partícula:5 µm
- Tamaño de poro:85 Å
- Longitud:250 mm
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:ISO 14001:2015 (530840 UM15)
- Referencia:76402-044
- Phase:Silica
Avantor® Partisil® was one of the first commercially available irregular silicas.
- Irregular porous silica
- 5 and 10 µm particle sizes
- Hichrom high efficiency
A large surface area gives it a high loading capacity. Avantor® Partisil® ion-exchange materials are widely referenced and remain one of the most popular choices for analysts.