Avantor® Partisil C8, HPLC Columns
Supplier: Avantor
Acerca de este artículo
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
- Easy to use
- Irregular porous silica
- Stainless steel column hardware
Avantor® Partisil® C8 is recommended for ion-pair chromatography.
: * Use with freestanding holder HI-161 and column coupler HI-081, please see 'HPLC Guard Column Holders and Accessories'.
2 Options Available Below
Product Details & Documents
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
- Easy to use
- Irregular porous silica
- Stainless steel column hardware
Avantor® Partisil® C8 is recommended for ion-pair chromatography.
: * Use with freestanding holder HI-161 and column coupler HI-081, please see 'HPLC Guard Column Holders and Accessories'.
Specifications for Product Family
Especificaciones
- Catalog No:
- 76402-062
- 76402-088
- 76398-238
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Especificaciones
Product Family Options
Analytical Columns
Información del producto
- PhasePore sizeParticle SizeØ int.LengthAvailabilityPrice
- Supplier Number: P5C8-100AEspecificaciones:
Low Manufacturing Impact More
Specifications
Cat. No.76402-062Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - C8, endcapped85 Å5 µm4.60 mm250 mmDiscontinued-Supplier Number: -Especificaciones:
Low Manufacturing Impact More
Specifications
Cat. No.76402-088Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Supplier Number: 4229-001Especificaciones:
Low Manufacturing Impact More
Specifications
Cat. No.76398-238Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact