Zorbax CN, HPLC Columns
CN, 70 Å, 5 µm, 4.00 mm, 250 mm
Catalog # 76402-522
Supplier: Avantor
Número de proveedor #: ZCN-250AF
Jump to:
Product Details & Documents
Zorbax spherical particles are manufactured from small extremely uniform colloidal silica micro beads which are agglutinated in a Du Pont patented organic polymerization process.
- Spherical porous silica
- High efficiency
- Narrow particles and pore size distribution
- Robust with good batch to batch reproducibility
The organic polymer is removed by a controlled sintering process to yield mechanically stable silica particles.
Especificaciones
- Ø int.:4.00 mm
- Particle Size:5 µm
- Pore size:70 Å
- Length:250 mm
- Phase:CN
- Carbon load:7%
- Cat. No.:76402-522
- Environmental certification:ISO 14001:2015 (530840 UM15)
- Endcapped:Yes
- Surface area:330 m²/g
- Environmentally Preferable:Low Manufacturing Impact