Avantor® Hichrom, Frits for Columns
Catalog # 76399-046
Supplier: Avantor
Specifications
- Porosidad:2.0 µm
- Descripción:Frits for 2.1 mm I.D. columns, 6.35 mm column Ø
- Frit Material:Stainless steel, PEEK coated
- Frit Thickness:1.575 mm
- Referencia:76399-046
Specifications
About this item
Frit made of 316 grade stainless steel.
- Helps in controlling porosity
A 2 μm porosity frit is recommended for retention of 5 to 20 μm packing material and 0.5 μm porosity frit is recommended for 3 to 4 μm packing materials.
Stainless steel with encapsulated PEEK used to prevent sample and eluent from entering dead volume space between the end fitting and also with the top of the column. This results in improved peak symmetry with 1.0 to 3.2 mm Int.Ø columns.