Avantor® Alltima HP C18, HPLC Columns
Catalog # 75797-386
Supplier: Avantor
Specifications
- Formato de columna:Analytical
- Ø int.:4.60 mm
- Tamaño de partícula:5 µm
- Descripción:Column
- Longitud:250 mm
- Extremo tapado:Yes
- Código USP:L1
- Referencia:75797-386
- Packing Material:Silica
- Phase:C18
- Nº de proveedor:87680
Specifications
About this item
Avantor® Alltima HP columns offer a range of different phase chemistries based on high purity silica. The Avantor® Alltima HP product family combines the selectivity and performance needed to overcome the most challenging separation needs.
- Excellent column stability
- Low to no detectable column bleed
- pH stability from 1 to 10
- Classic reversed-phase retention and selectivity
- Multiple selectivity options
The low column bleed makes these columns ideal for microbore applications.
Column hardware: Stainless steel