WHEATON® Rubber Stoppers, DWK Life Sciences
Mouth with ID×OD 15×30 mm, Snap-on, Gray, Bromobutyl
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Supplier #: W224100-342
WHEATON® Rubber Stoppers, DWK Life Sciences
Catalog # 10796-550
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Jump to:
Product Details & Documents
Clean and precise pharmaceutical grade rubber stoppers for research/parenteral drug applications.
- Variety of styles, sizes and rubber formulations
- Components manufactured to yield exceptional dimensional stability
- Straight plug stoppers provide maximum sealing for vials with straight-wall glass finish
- Snap-on style provides maximum sealing for vials with a blow-back glass finish
- Sleeve style stoppers feature a fold-over skirt for sealing in serum finish vials without a crimp closure
- Autoclavable
WHEATON® straight plug stoppers are manufactured from the highest quality raw materials.
Documents
Specifications
- Color:Gray
- Description:Snap-on
- Material:Bromobutyl
- Fits:Mouth with ID×OD 15×30 mm