Visual Scientifics Photogate System, Eisco Scientific
470317-428
- Descripción:Visual scientifics photogate system
- Referencia:470317-428
A complete photogate system for use with Visual Scientifics systems - or any lab experiment requiring photogates.
- Instruction manual included
- Platform neutral - works with any physics apparatus
This photogate system is an accessory for use with Visual Scientifics experiment kits, but it may be used for many other applications as well. This tool has a built in stopwatch, and features four function modes: gate, pulse, pendulum and frequency.
: Please note that the Visual Scientifics experiment kits and components (including the magnetic base) are not included with the photogate system.
: The system includes photogate timer, timer power cord, 2 photogates, 2 photogate cables, 2 support poles, 2 magnetic pole bases, and 2 sliding pole supports.