Avantor® Partisil SCX, HPLC Columns, 10 µm
76382-668
: Avantor
- Formato de columna:Analytical
- Ø int.:4.60 mm
- Tamaño de partícula:10 µm
- Tamaño de poro:85 Å
- Descripción:PARTISIL analytical HPLC columns with WCS hardware
- Longitud:250 mm
- Referencia:76382-668
- Phase:SCX
Avantor® Partisil® was one of the first commercially available irregular silicas. A large surface area gives it a high loading capacity.
- Irregular porous silica
- 5 and 10 µm particle sizes
- Hichrom high efficiency
- Stainless steel column hardware
Avantor® Partisil® ion-exchange materials are widely referenced and remain one of the most popular choices for analysts. Avantor® Partisil® SCX is a strong cation-exchange phase based on benzenesulphonic acid groups
Ideal for nucleosides, amino acids, polyamines, drugs and other cationic species.