Avantor® ACE® Excel® C18-AR, Analytical HPLC Columns, 3 µm
: Avantor
The Avantor® ACE® Excel® C18-AR phase utilizes a specially developed ligand combining a C18 chain with integral phenyl functionality, thus combining the benefits of both C18 and phenyl characteristics into a single phase. This can be used for ‘standard’ C18 column separations, but is additionally recommended for separations that involve compounds containing aromatic functionality.
- Ultra high purity, base deactivated silica with exceptional reproducibility
- Ideal for a wide range of analytes differing in hydrophobicity
- Wide range of particle sizes (1.7 to 5 µm) and complementary phases
- Ultra low bleed to ensure LC/MS compatibility
- Method validation kits contain three different batches of the same phase - available on request
These columns are made of stainless steel.
- Catalog No:
- 76393-712
- 76382-642
- 76393-706
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carga de carbono:
- 15.5%
- 15.5%
- 15.5%
- Extremo tapado:
- Yes
- Yes
- Yes
- Presión máx.:
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- 1000 bar (14504 psi)
- Rango de pH:
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- Código USP:
- L1
- L1
- L1
- Certificación ambiental:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Guard holders and accessories
Frequently Bought Together