Avantor® ACE® Excel® SuperC18, Analytical, HPLC/UHPLC Columns, 3 µm
Catalog # 76393-146
Supplier: Avantor
Specifications
- Formato de columna:Analytical
- Ø int.:2.10 mm
- Tamaño de partícula:3 µm
- Tamaño de poro:90 Å
- Presión máx.:1000 bar (14504 psi)
- Longitud:150 mm
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:ISO 14001:2015 (530840 UM15)
- Extremo tapado:Yes
- Código USP:L1
- Rango de pH:1.5 - 11.5
- Surface area:400 m²/g
- Carga de carbono:14.8%
- Referencia:76393-146
- Phase:SuperC18™
Specifications
About this item
Avantor® ACE® Excel® SuperC18™ columns provide high stability across an extended pH range. These columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.
- Ultra-inert for maximum performance and reproducibility
- Ultra-low bleed for improved LC/MS compatibility
- Wide range of particle sizes (1.7 to 10 µm) and complementary phases
These columns are made of stainless steel.