Avantor® ACE® Excel® C18, Analytical UHPLC Columns, 3 µm
Catalog # 76393-128
Proveedor: Avantor
Especificaciones
- Formato de columna:Analytical
- Ø int.:4.60 mm
- Tamaño de partícula:3 µm
- Tamaño de poro:100 Å
- Presión máx.:1000 bar (14504 psi)
- Longitud:100 mm
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:ISO 14001:2015 (530840 UM15)
- Extremo tapado:Yes
- Código USP:L1
- Rango de pH:2 - 8
- Surface area:300 m²/g
- Carga de carbono:15.5%
- Referencia:76393-128
- Phase:C18
Especificaciones
Acerca de este artículo
Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
- Ultra-high purity, base deactivated silica with exceptional reproducibility
- Ideal for a wide range of analytes differing in hydrophobicity
- Wide range of particle sizes (1.7 to 5 µm) and complementary phases
- Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.