Orders LinkContactUsLinkComponent
Avantor® ACE® Excel® C18-AR, HPLC Columns, 2 µm
   Opciones sostenibles disponibles
Supplier:   Avantor
Avantor® ACE® Excel® C18-AR, HPLC Columns, 2 µm
Supplier:  Avantor
   Opciones sostenibles disponibles
Certificates
Avantor® ACE® Excel® C18-AR, HPLC Columns, 2 µm
3 Options Available Below
Product Details & Documents
The Avantor® ACE® Excel® C18-AR phase utilizes a specially developed ligand combining a C18 chain with integral phenyl functionality, thus combining the benefits of both C18 and phenyl characteristics into a single phase. This can be used for 'standard' C18 column separations but is additionally recommended for separations that involve compounds containing aromatic functionality.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1.7 to 5 µm) and complementary phases
  • Ultra-low bleed to ensure LC/MS compatibility
  • Method validation kits contain three different batches of the same phase - available on request
These columns are made of stainless steel.
Specifications for Product Family
Especificaciones
  • Catalog No:
  • 76382-622
  • 76382-626
  • 76393-028
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Carbon load:
  • 15.5%
  • 15.5%
  • 15.5%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Max. pressure:
  • 1000 bar (14504 psi)
  • 1000 bar (14504 psi)
  • 1000 bar (14504 psi)
  • pH range:
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • Surface area:
  • 300 m²/g
  • 300 m²/g
  • 300 m²/g
  • USP code:
  • L1
  • L1
  • L1
  • Environmental certification:
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
  • ISO 14001:2015 (530840 UM15)
Especificaciones
Product Family Options
Información del producto
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • Analytical
    C18-AR
    100 Å
    2 µm
    2.10 mm
    50 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $11,059.98
    $11,059.98
    Each (1UN)
       $11,059.98
    Supplier Number: EXL-109-0502U
    Especificaciones:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    76382-622
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    15.5%
    Endcapped
    Yes
    Max. pressure
    1000 bar (14504 psi)
    pH range
    2 - 8
    Surface area
    300 m²/g
    USP code
    L1
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Analytical
    C18-AR
    100 Å
    2 µm
    2.10 mm
    100 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $13,022.88
    $13,022.88
    Each (1UN)
       $13,022.88
    Supplier Number: EXL-109-1002U
    Especificaciones:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    76382-626
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    15.5%
    Endcapped
    Yes
    Max. pressure
    1000 bar (14504 psi)
    pH range
    2 - 8
    Surface area
    300 m²/g
    USP code
    L1
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
  • Analytical
    C18-AR
    100 Å
    2 µm
    4.60 mm
    150 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $17,021.75
    $17,021.75
    Each (1UN)
       $17,021.75
    Supplier Number: EXL-109-1546U
    Especificaciones:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    76393-028
    Environmentally Preferable
    Low Manufacturing Impact
    Carbon load
    15.5%
    Endcapped
    Yes
    Max. pressure
    1000 bar (14504 psi)
    pH range
    2 - 8
    Surface area
    300 m²/g
    USP code
    L1
    Environmental certification
    ISO 14001:2015 (530840 UM15)
      Low Manufacturing Impact
Guard holders and accessories
Información del producto
  • Catalog #
    Description
    Availability
    Price
  • UHPLC guard cartridge holder for 2.1 to 4.6 mm Int.Ø. columns
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $4,713.94
    $4,713.94
    Each (1UN)
       $4,713.94