VAI® SMS Garments, Veltek
The SMS garment line is constructed with a high quality spunbond-meltblown-spunbond (SMS) non-woven polypropylene fabric that provides excellent breathability while maintaining protection.
- Highly breathable fabric releases heat while maintaining protection
- Garments feature elastic in high movement areas for ease of movement and appropriate fit
- 100% bound seam construction for increased durability and reduced particle permeability
- Individually packaged in easy to tear bags with two liner bags per case
- All packaging is labled with lot number and expiration date
All SMS coveralls are equipped with elastic wrists and ankles, along with a zipper closure. All SMS frocks feature elastic wrists and a zipper closure.
: All garments are supplied with a one-year warranty.
Frequently Bought Together