Distillation Adapter, 75°, with BEVEL-SEAL™, DWK Life Sciences
Catálogo # KT274960-0000
Proveedor: DWK LIFE SCIENCES S. DE R.L. DE MX
Especificaciones
- Anchura:50 mm
- Descripción:Distillation adapter, 75°, with BEVEL-SEAL™
- Altura:75 mm
- Referencia:KT274960-0000
- ST Joint Size:14/20
- Nº de proveedor:274960-0000
Especificaciones
Acerca de este artículo
75° Connecting distillation adapter with BEVEL-SEAL™ used in distillation assemblies where adjustable immersion depth of the thermometer is desired.
- Threaded top BEVEL-SEAL™ connection for plain stem thermometers
- Vacuum-tight seal is achieved with the open top compression cap and FKM O-ring
BEVEL-SEAL™ cap (410119-1307) is suitable for use to 200 °C and thread size is modified GPI 13-425 and FKM o-ring is size 010.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.