Chromatography Paper
Chromatography paper, pack of 50 strips
Catalog # 470320-990
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: CHRP050
Jump to:
Product Details & Documents
Separate the different colors of a solution.
- Dimension: 12.5×1.75 cm
Chromatography paper is used to separate colored chemicals. This is done by taking advantage of the different rates of migration across paper.
Packaging: Sold as a pack of 50 strips.
Especificaciones
- Description:Chromatography paper, pack of 50 strips