Deluxe Support Stand
Deluxe support stand, 5×8", ⅜", 20"
470320-982
: SSB5X8-J
Support stand that ensures the stand stays firmly in place.
- Built in torqueing rod
- Thick mounting hole for a more secure connection
Cast iron bases with gray enamel finish. The base is 3 mm thick with a welded plate on the mounting hole. This gives the support rod 10 mm of screw support, which is three times thicker than other traditional stands. The support rod includes an 80 mm long rod to provide more torque when securing the rod to the base plate.
: Includes a removable, zinc-plated steel rod.
- Rod Length:20"
- Description:Deluxe support stand
- Base Dimensions:5×8"
- Rod Diameter:⅜"