Clear Plastic Overflow Can
Clear plastic overflow can
Catalog # 470320-742
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: OFLCN2
Jump to:
Product Details & Documents
This transparent overflow can is graduated to 125 ml and includes an attached spout.
- Clear design
Made of clear acrylic, the can measures 5.5 cm I.D.×7 cm high with a wall thickness of 3 mm.
Especificaciones
- Description:Clear plastic overflow can