Mini Projectile Launcher
Mini projectile launcher
Catalog # 470320-736
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: MPL001
Jump to:
Product Details & Documents
Investigate horizontal and vertical motion.
- Projectile range of two meters
- Multiple launch velocities
This simple but precise launcher projects 16 mm steel balls at ranges suitable for use on the bench top or from the bench to the floor. A protractor and plumb line make it easy to set any required launch angle from 45° downwards to straight up.
Three different spring tensions give three repeatable launch speeds (ranges of 1 m, 1.5 m and 2 m) and a magnetic piston ensures that the ball does not touch the side of the barrel and launches straight every time.
A cable release gives a smooth triggering to prevent jerking. The unit can stand on its own or be fixed to the bench with a clamp.
Documents
Especificaciones
- Description:Mini projectile launcher