KIMBLE® Recovery Flasks with Square Joint Bead, DWK Life Sciences
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificates
About this item
Single neck flask with a standard taper outer joint designed for easy recovery of reaction products.
- Square bead joints provide superior clamping to standard joint beads
- Total contents are accessible with a spatula or policeman
- Lower portion fits standard heating mantles
- Manufactured from 33 expansion, low extractable borosilicate glass
Even wall thickness and minimum runout make these flasks ideal for use with rotary evaporators.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Frequently Bought Together