Water Molecule Model Set, Eisco Scientific
Single water molecule set
Catalog # 470309-118
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: CHMOLH2O
Jump to:
Product Details & Documents
Five part set to demonstrate the structure of a single water molecule.
- Includes (2) white Hydrogen balls, (1) red Oxygen ball and (2) gray molecular bonds
- Can be used as interchangeable parts for inorganic and organic chemistry molecular model kits
- Excellent visual learning tool for students
This single water molecule set is great for demonstrations or for use as additional/replacement parts for molecular model kits. Assembled, this molecular model measures 2.6"
Delivery information: Kit Includes (2) 14 mm white Hydrogen balls, (1) 19 mm red Oxygen ball, and (2) gray molecular bonds measuring 30 mm each.
Especificaciones
- Description:Single water molecule set