Universal 2 Prong Clamp with Bosshead, Eisco Scientific
Catálogo # 470231-448
Proveedor: EISCO SCIENTIFIC LLC
Especificaciones
- Descripción:Universal 2 prong clamp with bosshead
- Referencia:470231-448
Especificaciones
Acerca de este artículo
A versatile clamp to securely grip a variety of cylindrically shaped glass and labware.
- Manufactured with stamped steel
- Three adjustable thumb screws
- Integrated boss head
- Features one flat angled jaw and one V shaped jaw
- Jaws are coated in black vinyl
- Provides a secure grip for all cylindrical shaped objects
High quality, reliable universal steel clamp for holding and securing a variety of cylindrically shaped labware. Features include an integrated boss head, thumb screws, and vinyl coated jaws.