Orders LinkContactUsLinkComponent
Environmental Testing Kit
Environmental Testing Kit
Environmental Testing Kit
Catalog # 470317-456
Supplier:   ALDON CORP SE
Supplier #:  IS9799
Environmental Testing Kit
Catalog # 470317-456
Supplier:  ALDON CORP SE
Environmental Testing Kit
Environmental Testing Kit
Environmental Testing Kit
Price
$2,010.05
Log in for availability
Kit de 1
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Learn how environmental scientists study pollution by testing air and water samples from your local area.
  • Kit includes nine different tests to determine the air and water quality of samples collected from your community
  • There are enough materials for 15 groups of students
  • Teacher's Manual and Student Study Guide copymaster are included
Investigate your water quality by testing for dissolved oxygen levels, water hardness, pH, and phosphate and chloride content. Then test for particulate and chemical pollutants in the air, measure smoke density, and learn how excess carbon dioxide in air can affect the environment.
Aligned to the Next Generation Science Standards (NGSS)*.
Disciplinary Core Ideas: ESS3.A; ESS3.C
Performance Expectations: 4-ESS3-1; MS-ESS3-3; HS-ESS3-4
Cross Cutting Concepts: Cause and Effect; Interdependence of Science, Engineering, and Technology; Influence of Science, Engineering, and Technology on Society and the Natural World; Influence of Engineering, Technology, and Science on Society and the Natural World
Engineering Practices: Obtaining, Evaluating, and Communicating Information; Constructing Explanations and Designing Solutions
 : Kit includes: 25 ml Methylene Blue, 0.1%, 25 ml EDTA Solution, 0.015 M, 25 ml Silver Nitrate, 0.014 M, 100 ml Bromothymol Blue, 0.04%, 3 Hardness Powder Pillows, (3) Phosphate Powder Pillows, (3) Chloride Powder Pillows, (1) glass bottle, 4oz. with cap, (1) package pH strips, (1) plastic vial with cap, (1) medicine cup, (2) cups, 10 oz., (2) magnifiers, (4) clear labels, (1) nylon fabric, (2) rubber bands, (1) Smoke Density comparator, (1) Phosphate indicator, and (1) syringe.
Specifications
  • Description:
    Environmental Testing Kit
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.