Rubbermaid® Commercial Clean Room Maintenance Mop Heads
Catálogo # 66103-130
Proveedor: ESSENDANT LLC FKA LAGASSE
Especificaciones
- Referencia:66103-130
- Nº de proveedor:RCP T300
- Headband Size:5"
- Mop Head Size:Medium
Especificaciones
Acerca de este artículo
Sturdy, looped-end tailbanded floor mop for industrial applications, including pharmaceutical labs, paint booths, and more.
- Made of launderable, pre-shrunk rayon/polyester fabric that will not fray or lint
- Headband color: Green
- Mop head color: White