Orders LinkContactUsLinkComponent
Wringers, Contec®
Wringers, Contec®
Wringers, Contec®
Supplier:  CONTEC, INC. MX
Certificates
Wringers, Contec®
Wringers, Contec®
4 Options Available Below
Product Details & Documents
A variety of wringers designed for use with curved and flat mops.
  • Stainless steel and polypropylene wringers for a variety of Contec® bucket systems
  • Can be used with curved, flat, and string mops
  • Fully autoclavable
These wringers fit on Contec® 3.9 gallon (15 l), 5 gallon (20 l) , and 6.5 gallon (25 l) bucket systems. All wringers are compatible with a wide variety of disinfectants.
Product Family Options
Product Information
  • Material
    For Use With
    Availability
    Price
  • 89198-850
    Downpress Flat Mop Wringer
    Downpress Flat Mop Wringer
    Stainless steel
    VertiKlean® MAX™ and QuickTask™ Mops
    Log in for availability
    Log in for availability
    Case (1UN)
    Case (1UN) - $31,251.21
    $31,251.21
    Case (1UN)
       $31,251.21
    Supplier Number: 2756
  • 76199-888
    Curved Mop Wringer
    Curved Mop Wringer
    Stainless steel
    EasyCurve® Mops (15 l (3.9 gal.) and 25 l (6.5 gal.) Buckets)
    Log in for availability
    Log in for availability
    Case (1UN)
    Case (1UN) - $7,683.07
    $7,683.07
    Case (1UN)
       $7,683.07
    Supplier Number: 2690
  • Flat Mop Wringer
    PP
    VertiKlean® MAX™ and QuickTask™ Mops (25 l (6.5 gal.) Buckets)
    Log in for availability
    Log in for availability
    Case (1UN)
    Case (1UN) - $1,818.75
    $1,818.75
    Case (1UN)
       $1,818.75
    Supplier Number: 2738
  • Flat Mop Wringer
    Stainless steel
    VertiKlean® MAX™ and QuickTask™ Mops (15 l (3.9 gal.) and 25 l (6.5 gal.) Buckets)
    Log in for availability
    Log in for availability
    Case (1UN)
    Case (1UN) - $6,892.09
    $6,892.09
    Case (1UN)
       $6,892.09
    Supplier Number: 2689