QuickConnect™ Adaptor, Contec®
QuickConnect™ Adaptor, 13.6×2.3 cm (5.35×0.9"), Stainless steel
Supplier: CONTEC, INC. MX
Supplier #: 2758
Jump to:
Product Details & Documents
Stainless steel adaptor for use with QuickTask™ mops.
- Autoclavable at 121 °C for 30 minutes
The QuickConnect™ Adaptor allows users to connect any QuickConnect™ handle to the QuickTask™ polypropylene mop frame.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:QuickConnect™ Adaptor
- Material:Stainless steel
- Size:13.6×2.3 cm (5.35×0.9")