Swift M3600 Series Compound Microscopes
Catálogo # 470317-656
Proveedor: Swift Instruments
Especificaciones
- Modelo:M3604C-4
- Descripción:4×, 10×, 400×, 1000× with 1.25 Abbe Condenser and Built-In Mech. Stage
- Referencia:470317-656
- Illumination:Cordless LED
Especificaciones
Acerca de este artículo
This microscope series is perfect for high school students.
- Lead-free objectives
- Coaxial coarse and fine focus
- Variable LED illumination
- Dimmer switch
- Built-in camera port
Now with coaxial focusing and upgraded LED illumination, the M3600 Series continues the Swift tradition for innovation, quality, and student-proof features.